Specifications
- Description:Larvae T-maze, zebrafish
- Material:Acrylic
- Environmentally Preferable:
- Cat. No.:76670-118
Specifications
About this item
The Zebrafish larvae T-Maze takes advantage of unique backlighting to allow for fine behavior task analysis in larvae.
- Clear acrylic construction
- Includes lid, chamber, and backlight
Similar to the drosophila mazes, this apparatus comes with a start lane, bidirectional swimming pools, and a unique backlight for easy video tracking. An easy-to-use cover seals the pools and watertight chambers ensure a long service life.