Geno/Grinder® HG-600, 2010 High-Throughput Homogenizer
Catalog # MSPP-EW0457684
Supplier: Cole Parmer
Specifications
- Description:Geno/Grinder® 2010 high throughput homogenizer
- Dimensions:35.6×71.1×57.2 cm (14.0×28.0×22.5")
- Environmentally Preferable:
- Cat. No.:MSPP-EW0457684
- Electrical:230 VAC, 50 Hz
- Speed range:Adjustable from 500 - 1750 strokes/minute
- Sample volume:Accommodates full range of sample vials from 2 - 50 ml
- Weight:48 kg (106 lbs.)
Specifications
About this item
Accommodates full range of sample vials from 2 to 50 ml. Fast and efficient extraction of nucleic acids, proteins, and other molecules. Samples can be processes with or without reagents in approximately 60 seconds or less.
- Double-locking lid provides safe operation
- Sound insulation and suspension system ensure quiet, vibration-free operation
- Fully adjustable clamp allows simple front loading of vials or microplates
- Interior LED lighting and large window improve sample visibility
- Fully programmable control screen tilts for optimal viewing angle
- Adjustable clamp speed from 500 to 1700 strokes/min
- Clamp provides 3.2 cm (1.25") vertical movement, for 230 VAC operation
- Three year warranty
The password-protected, touch screen control panel enables you to program run time, rate, cycles, and pause time. The control panel can be tilted for better viewing. Up to 500 protocols can be easily labeled, saved, recalled, and exported via USB.
Ordering information: Includes: Adjustable clamp assembly and two-prong grounded plug with cord.