Anti-mCherry Mouse Monoclonal Antibody [clone: 1C51]
Mouse monoclonal (1C51) antibody to mCherry for WB, ICC/IF and IHC.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:2000, ICC/IF: 1:500, IHC: 1:500
- Host: Mouse
- Clonality: Monoclonal
- Isotype: IgG2a
- Conjugate: Unconjugat
Type: Primary
Antigen: mCherry
Clonality: Monoclonal
Clone: 1C51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
- Catalog No:
- 77068-396
- 77072-978
- Antigen Symbol:
- mCherry
- mCherry
- Antigen Name:
- mCherry
- mCherry
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 1C51
- 1C51
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# D1MPT3
- UniprotID# D1MPT3
- Isotype:
- IgG2a
- IgG2a
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length mCherry, expressed in and purified from E. coli
- Recombinant full-length mCherry, expressed in and purified from E. coli
- Sequence:
- MVSKGEEDNMAIIKEFMRFKVHMEGSVNGHEFEIEGEGEGRPYEGTQTAKLKVTKGGPLPFAWDILSPQFMYGSKAYVKHPADIPDYLKLSFPEGFKWERVMNFEDGGVVTVTQDSSLQDGEFIYKVKLRGTNFPSDGPVMQKKTMGWEASSERMYPEDGALKGEIKQRLKLKDGGHYDAEVKTTYKAKKPVQLPGAYNvNIKLDITSHNEDYTIVEQYERAEGRHSTGGMDELYK
- MVSKGEEDNMAIIKEFMRFKVHMEGSVNGHEFEIEGEGEGRPYEGTQTAKLKVTKGGPLPFAWDILSPQFMYGSKAYVKHPADIPDYLKLSFPEGFKWERVMNFEDGGVVTVTQDSSLQDGEFIYKVKLRGTNFPSDGPVMQKKTMGWEASSERMYPEDGALKGEIKQRLKLKDGGHYDAEVKTTYKAKKPVQLPGAYNvNIKLDITSHNEDYTIVEQYERAEGRHSTGGMDELYK
- Form:
- Liquid
- Liquid
- Molecular Weight:
- ~28 kDa
- ~28 kDa
- Purification:
- Immunogen affinity purification.
- Immunogen affinity purification.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF, IHC
- ICC/IF, IHC