Anti-GST Mouse Monoclonal Antibody [clone: GST.B6]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal (GST.B6) antibody to GST Tag for Dot, ELISA, IP, IS and WB.
- Validated Applications: Dot, ELISA, IP, IS, WB
- Host: Mouse
- Clonality: Monoclonal
- Isotype: IgG2a
- Conjugate: Unconjugat
Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone: GST.B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Specifications
- Catalog No:
- 77065-250
- 77075-414
- Antigen Symbol:
- GST
- GST
- Antigen Name:
- Glutathione S Transferase
- Glutathione S Transferase
- Antigen Synonyms:
- Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|GST class alpha 2|GST|GST class alpha 1|GST gamma|Glutathione S transferase A1|Glutathione S transferase A2|GSTA1
- Glutathione S-transferase class-mu 26 kDa isozyme|GST epsilon|GST class alpha 2|GST|GST class alpha 1|GST gamma|Glutathione S transferase A1|Glutathione S transferase A2|GSTA1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- GST.B6
- GST.B6
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- GST protein
- GST protein
- Form:
- Liquid
- Liquid
- Purification:
- Protein A affinity chromatography from mouse ascites fluid.
- Protein A affinity chromatography from mouse ascites fluid.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Supplied in 10 mM Phosphate buffered saline, pH 7.20, with 0.05% Sodium Azide
- Supplied in 10 mM Phosphate buffered saline, pH 7.20, with 0.05% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested Applications:
- Dot Blot
- Dot Blot