Anti-Cytochrome P450 2D10 Goat Polyclonal Antibody
Catalog # 76950-778
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CP2DA_RAT
- Antigen Symbol:Cytochrome P450 2D10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Cytochrome P450-DB5|Cytochrome P450-CMF1B|Debrisoquine 4-hydroxylase|CYPIID10|Cytochrome P450 2D10|Cyp2d10
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:CDAFTAQNGRSINPK
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Rat Cyp2d5 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:76950-778
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to Cytochrome P450 2D10 for ELISA and WB with samples derived from Rat.
- Validated Applications: ELISA, W
Type: Primary
Antigen: Cytochrome P450 2D10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat