Anti-TRIM11 Goat Polyclonal Antibody
77069-126
- Antibody Type:Primary
- Antigen Name:BIA 1
- Antigen Symbol:TRIM11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:E3 ubiquitin-protein ligase TRIM11|BIA1|Protein BIA1|RNF92|E3 ubiquitin protein ligase TRIM11|RING type E3 ubiquitin transferase TRIM11|RING finger protein 92|TRI11_HUMAN|RNF 92
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:CRPKGGSGDTLAPQ
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human TRIM11 (C terminal)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77069-126
- Packaging:Plastic vial
Goat polyclonal antibody to TRIM11 for ELISA and IHC with samples derived from Human.
- Validated Applications: ELISA, IH
Type: Primary
Antigen: TRIM11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human