Anti-IGFBP1 Rabbit Monoclonal Antibody [clone: ARC0671]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC0671] antibody to IGFBP1 for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: IGFBP1
Clonality: Monoclonal
Clone: ARC0671
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77209-162
- Antigen Symbol:
- IGFBP1
- Antigen Name:
- AFBP
- Antigen Synonyms:
- IBP 1|Binding protein 28|hIGFBP1|hIGFBP 1|Alpha pregnancy associated endometrial globulin|Binding protein 25|Amniotic fluid binding protein|Growth hormone independent binding protein|Binding protein 26
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC0671
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P08833
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 160-259 of human IGFBP1 (P08833)
- Sequence:
- GSKALHVTNIKKWKEPCRIELYRVVESLAKAQETSGEEISKFYLPNCNKNGFYHSRQCETSMDGEAGLCWCVYPWNGKRIPGSPEIRGDPNCQIYFNVQN
- Form:
- Liquid
- Molecular Weight:
- 30 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C