Anti-Artemis Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Artemis for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: Artemis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77217-170
- Antigen Symbol:
- Artemis
- Antigen Name:
- A SCID
- Antigen Synonyms:
- DCLRE1C|Artemis protein|A SCID protein|DCLRE1C protein|Artemis|ASCID|DCLRE1C DNA cross link repair 1C|DCR1C_HUMAN|DCLREC1C
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q96SD1
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human DCLRE1C (NP_001029027.1)
- Sequence:
- MSSFEGQMAEYPTISIDRFDRENLRARAYFLSHCHKDHMKGLRAPTLKRRLECSLKVYLYCSPVTKELLLTSPKYRFWKKRIISIEIETPTQISLVDEASGEKEEIVVTLLPAGHCPGSVMFLFQGNNGTVLYTGDFRLAQGEAARMELLHSGGRVKDIQSVYLDTTFCDPRFYQIPSREECLSGVLELVRSWITRSPYHVVWLNCKAAYGYEYLFTNLSEELGVQVHVNKLDMFRNMPEILHHLTTDRNTQIHACRHPKAEEYFQWSKLPCGITSRNRIPLHIISIKPSTMWFGERSRK
- Form:
- Liquid
- Molecular Weight:
- 90 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C