Anti-Sodium Iodide Symporter Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to Sodium Iodide Symporter for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:20 to 1:5
Type: Primary
Antigen: Sodium Iodide Symporter
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 76861-180
- Antigen Symbol:
- Sodium Iodide Symporter
- Antigen Name:
- Na(+)/I(-) cotransporter
- Antigen Synonyms:
- NIS|Sodium Iodide Symporter|Na+/I- cotransporter|SC5A5_HUMAN|Na(+)/I(-) symporter|Slc5a5|Na+/I-symporter|Sodium-iodide symporter|Sodium/iodide cotransporter
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q92911
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 544-643 of human SLC5A5 (NP_000444.1)
- Sequence:
- ALISCLTGPTKRSTLAPGLLWWDLARQTASVAPKEEVAILDDNLVKGPEELPTGNKKPPGFLPTNEDRLFFLGQKELEGAGSWTPCVGHDGGRDQQETNL
- Form:
- Liquid
- Molecular Weight:
- 68 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF