Anti-Lactate Dehydrogenase B/LDH-B Rabbit Monoclonal Antibody [clone: ARC1217]
Supplier: Avantor
Certificates
About this item
Rabbit monoclonal [ARC1217] antibody to Lactate Dehydrogenase B/LDH-B for WB, IHC and IP with samples derived from Human and Rat.
- Validated Applications: WB, IHC, IP
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, IP: 1:500 to 1:1,000
Type: Primary
Antigen: Lactate Dehydrogenase B / LDH-B
Clonality: Monoclonal
Clone: ARC1217
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat