Anti-ENT2 Rabbit Monoclonal Antibody [clone: ARC2234]
Rabbit monoclonal [ARC2234] antibody to ENT2 for IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ICC/IF
- Recommended Dilutions: IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: ENT2
Clonality: Monoclonal
Clone: ARC2234
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77224-236
- Antibody Type:
- Primary