Anti-AKAP14 Rabbit Monoclonal Antibody [clone: ARC2344]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2344] antibody to AKAP14 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: AKAP14
Clonality: Monoclonal
Clone: ARC2344
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77221-010
- Antigen Symbol:
- AKAP14
- Antigen Name:
- A kinase (PRKA) anchor protein 14
- Antigen Synonyms:
- AKAP28|A kinase anchor protein 14|PRKA14|Protein kinase A anchoring protein 14|A-kinase anchoring protein 28|A kinase anchor protein 28 kDa|AKAP14
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC2344
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q86UN6
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 70-150 of human AKAP14 (Q86UN6).
- Sequence:
- HGEFTVEKGLKQIDEYFSKCVSKKCWAHGVEFVERKDLIHSFLYIYYVHWSISTADLPVARISAGTYFTMKVSKTKPPDAP
- Form:
- Liquid
- Molecular Weight:
- 23 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C