Anti-GRK3 Rabbit Monoclonal Antibody [clone: ARC1458]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1458] antibody to GRK3 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: GRK3
Clonality: Monoclonal
Clone: ARC1458
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77219-650
- Antigen Symbol:
- GRK3
- Antigen Name:
- ADRBK2
- Antigen Synonyms:
- Beta-adrenergic receptor kinase 2|G protein coupled receptor kinase 3|Beta ARK 2|Adrenergic, beta, receptor kinase 2|BARK2|EC 2.7.11.15|Beta-ARK-2|Beta adrenergic receptor kinase 2|ARBK2_HUMAN
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1458
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# P35626
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 589-688 of human GRK3 (P35626).
- Sequence:
- EWRGEGESRQNLLTMEQILSVEETQIKDKKCILFRIKGGKQFVLQCESDPEFVQWKKELNETFKEAQRLLRRAPKFLNKPRSGTVELPKPSLCHRNSNGL
- Form:
- Liquid
- Molecular Weight:
- 80 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C