About this item
Rabbit monoclonal [ARC0829] antibody to KMT1B/SUV39H2 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: KMT1B / SUV39H2
Clonality: Monoclonal
Clone: ARC0829
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77219-498
- Antigen Symbol:
- KMT1B / SUV39H2
- Antigen Name:
- FLJ23414
- Antigen Synonyms:
- Histone-lysine N-methyltransferase SUV39H2|Histone H3-K9 methyltransferase 2|Histone lysine N methyltransferase SUV39H2|H3-K9-HMTase 2|Histone lysine N methyltransferase H3 lysine 9 specific 2|H3 K9 HMTase 2|KMT1B|KMT1B / SUV39H2|Histone H3 K9 methyltransferase 2
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC0829
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9H5I1
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 311-410 of human KMT1B/SUV39H2 (Q9H5I1).
- Sequence:
- ESDEFTVDAARYGNVSHFVNHSCDPNLQVFNVFIDNLDTRLPRIALFSTRTINAGEELTFDYQMKGSGDISSDSIDHSPAKKRVRTVCKCGAVTCRGYLN
- Form:
- Liquid
- Molecular Weight:
- 47 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C