Anti-WDR79 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to WDR79 for WB with samples derived from Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: WDR79
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77218-234
- Antigen Symbol:
- WDR79
- Antigen Name:
- DKCB3
- Antigen Synonyms:
- WDR79|WD repeat containing antisense to TP53|FLJ10385|TCAB1|WAP53_HUMAN|Telomerase Cajal body protein 1|WD40 repeat-containing protein encoding RNA antisense to p53|WD repeat containing protein 79|WD repeat-containing protein 79
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9BUR4
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 62-160 of human WRAP53 (NP_060551.2).
- Sequence:
- AVSQELREGDPVSLSTPLETEFGSPSELSPRIEEQELSENTSLPAEEANGSLSEEEANGPELGSGKAMEDTSGEPAAEDEGDTAWNYSFSQLPRFLSGS
- Form:
- Liquid
- Molecular Weight:
- 72 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C