Anti-mGluR5 Mouse Monoclonal Antibody [clone: S75-3]
Catalog # 77172-666
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:mGluR5
- Clonality:Monoclonal
- Clone:S75-3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P31424
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~130 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b.
- Tested Applications:IHC, ICC/IF
- Purification:Protein G purification
- Cat. No.:77172-666
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [S75-3] antibody to mGluR5 for WB, IHC, ICC/IF and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF, IP
- Recommended Dilutions: WB: 1:1,000, IHC: 1:1,000, ICC/IF: 1:1,000
Type: Primary
Antigen: mGluR5
Clonality: Monoclonal
Clone: S75-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse;Rat