Anti-CREBBP Rabbit Polyclonal Antibody
Catalog # 77172-494
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:CBP
- Antigen Symbol:CREBBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q92793
- Antigen Synonyms:CREB binding protein|RTS|RSTS|CBP_HUMAN|KAT3A / CBP|Crebbp|Cyclic AMP responsive enhancer binding protein|KAT3A|CREB-binding protein
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:265 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide from the mid-protein of Human CBP (aa. 2100-2200).
- Tested Applications:IHC
- Purification:peptide. affinity purification
- Cat. No.:77172-494
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CREBBP for WB and IHC with samples derived from Human.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:1,000, IHC: 1:50
Type: Primary
Antigen: CREBBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human