To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:CACNA1G
- Antigen Symbol:CACNA1G
- Clonality:Monoclonal
- Clone:S178A-9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9WUT2
- Antigen Synonyms:KIAA1123|cav3 1c|Cav3 1|calcium channel voltage dependent T type alpha 1G subunit|CaV T1|calcium channel voltage dependent alpha 1G subunit|calcium channel voltage dependent T type alpha1G subunit|MGC117234|NBR13
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:>200 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 2052-2172 (cytoplasmic C-terminus) of mouse Cav3.1.
- Tested Applications:IHC, ICC/IF
- Purification:Protein G purification
- Cat. No.:77172-254
- Packaging:Plastic vial
Mouse monoclonal [S178A-9] antibody to CACNA1G for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100,
Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat