To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Symbol:Mouse IgG2c
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:500 µg
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied in 50 mM HEPES, pH 7.1, with 0.1 M Sodium Chloride, 1 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 0.2% BSA, and 0.09% Sodium Azide
- Storage Temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76833-558
- Packaging:Plastic vial
Specifications
About this item
Goat anti-mouse IgG2c (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Mouse.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:1,000 to 1:10,000, ICC: 1:100 to 1:1,000, IHC: 1:100 to 1:1,000, WB: 1:1,000 to 1:10,000
Type: Secondary
Antigen: Mouse IgG2c
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse