Anti-SARS-CoV-2 Nucleocapsid Protein Rabbit Recombinant Antibody [clone: RM420]
Catalog # 76834-424
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Symbol:SARS-CoV-2 Nucleocapsid Protein
- Clonality:Recombinant
- Clone:RM420
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:SARS-CoV-2
- Size:100 µg
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant SARS-CoV-2 Nucleocapsid protein, expressed in and purified from E. coli.
- Purification:Protein A chromatography
- Cat. No.:76834-424
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [RM420] antibody to SARS-CoV-2 Nucleocapsid Protein for ELISA with samples derived from SARS-CoV-2.
- Validated Applications: ELISA
Type: Primary
Antigen: SARS-CoV-2 Nucleocapsid Protein
Clonality: Recombinant
Clone: RM420
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: SARS-CoV-2