To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:antigen, prostate-specific
- Antigen Symbol:Prostate Specific Antigen
- Clonality:Recombinant
- Clone:RM323
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P07288
- Antigen Synonyms:Kallikrein 3|Kallikrein-3|Gamma seminoprotein|KLK 3|APS|Gamma-seminoprotein|hK3|Kallikrein related peptidase 3|KLK2A1
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Native PSA protein purified from human seminal fluid.
- Tested Applications:IHC
- Purification:Protein A chromatography
- Cat. No.:76834-620
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [RM323] antibody to Prostate Specific Antigen (Biotin) for IHC, WB and ELISA with samples derived from Human.
- Validated Applications: IHC, WB, ELISA
- Recommended Dilutions: IHC: 1:500 to 1:1,000, WB (non to reduced): 1:1,000 to 1:2,500, ELISA: 1:100 to 1:5,000
Type: Primary
Antigen: Prostate Specific Antigen
Clonality: Recombinant
Clone: RM323
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human