Order Entry
United States
ContactUsLinkComponent
Anti-GSK3 beta (phospho Ser9) Rabbit Recombinant Antibody [clone: RM453]
Anti-GSK3 beta (phospho Ser9) Rabbit Recombinant Antibody [clone: RM453]
Catalog # 76833-408
Supplier:  ANTIBODIES.COM LLC
Anti-GSK3 beta (phospho Ser9) Rabbit Recombinant Antibody [clone: RM453]
Catalog # 76833-408
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A290831-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Host:
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Cat. No.:
    76833-408

Specifications

About this item

Recombinant rabbit monoclonal [RM453] antibody to GSK3 beta (phospho Ser9) for WB with samples derived from Human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:1,000 to 1:2,000

Type: Primary
Antigen: GSK3 beta
Clonality: Recombinant
Clone: RM453
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human