Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-CD66c Mouse Monoclonal Antibody [clone: B6.2] (PE (Phycoerythrin))
Anti-CD66c Mouse Monoclonal Antibody [clone: B6.2] (PE (Phycoerythrin))
  76952-952
undefined
Anti-CD66c Mouse Monoclonal Antibody [clone: B6.2] (PE (Phycoerythrin))
  76952-952
 :  A285840-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Cat. No.:
    76952-952

 

 

Mouse monoclonal [B6.2] antibody to CD66c (PE) for Flow Cytometry with samples derived from Human.

  • Validated Applications: Flow Cytometry
  • Recommended Dilutions: Flow Cytometry: The reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD66c
Clonality: Monoclonal
Clone: B6.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human