Anti-MiTF Mouse Monoclonal Antibody [clone: PCRP-MITF-1D9]
76963-622
- Antibody Type:Primary
- Antigen Name:bHLHe32
- Antigen Symbol:MiTF
- Clonality:Monoclonal
- Clone:PCRP-MITF-1D9
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O75030
- Antigen Synonyms:Homolog of mouse microphthalmia|Microphthalmia, mouse, homolog of|Microphthalmia associated transcription factor|CMM8|Class E basic helix-loop-helix protein 32|MiTF|Microphthalmia-associated transcription factor|mitfa|Mi
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:52 - 56 kDa (doublet)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MiTF protein.
- Purification:Protein A/G chromatography
- Cat. No.:76963-622
- Packaging:Plastic vial
Mouse monoclonal [PCRP-MITF-1D9] antibody to MiTF for IP, IF, Flow Cytometry and WB with samples derived from Human.
- Validated Applications: IP, IF, Flow Cytometry, WB
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, IF: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml
Type: Primary
Antigen: MiTF
Clonality: Monoclonal
Clone: PCRP-MITF-1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human