To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cat. No.:76966-870
Mouse monoclonal [HNK-1 or Leu-7] antibody to CD57 for IF and IHC-P with samples derived from Human.
- Validated Applications: IF, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: HNK-1 or Leu-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human