Anti-Macrophage + Granulocyte Mouse Monoclonal Antibody [clone: BM-2]
Catalog # 76956-902
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:Macrophage + Granulocyte
- Antigen Symbol:Macrophage + Granulocyte
- Clonality:Monoclonal
- Clone:BM-2
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:183 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76956-902
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [BM-2] antibody to Macrophage + Granulocyte for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Macrophage + Granulocyte
Clonality: Monoclonal
Clone: BM-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human