Anti-pan HLA Class II Rabbit Recombinant Antibody [clone: HLA-Pan/8311R]
Catalog # 77180-592
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:pan HLA Class II
- Antigen Symbol:pan HLA Class II
- Clonality:Recombinant
- Clone:HLA-Pan/8311R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P04440 (HLA-DP), P01908, P01909, P01920 (HLA-DQ), P01903 (HLA-DR)
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:33 - 35 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Priess human B cell line.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-592
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [HLA-Pan/8311R] antibody to pan HLA Class II for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: pan HLA Class II
Clonality: Recombinant
Clone: HLA-Pan/8311R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human