Anti-Liver Arginase Rabbit Recombinant Antibody [clone: ARG1/8869R]
77180-190
- Antibody Type:Primary
- Antigen Name:A I
- Antigen Symbol:Liver Arginase
- Clonality:Recombinant
- Clone:ARG1/8869R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P05089
- Antigen Synonyms:Al|ARG 1|Arginase-1|Arginase 1|Arginase type I|ARGI1_HUMAN|Arginase, liver|Arginase liver|arg1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:35 - 38 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant human ARG1 protein fragment (aa1-200). The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77180-190
- Packaging:Plastic vial
Recombinant rabbit monoclonal [ARG1/8869R] antibody to Liver Arginase for WB and IHC-P with samples derived from Human.
- Validated Applications: WB, IHC-P
- Recommended Dilutions: WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Liver Arginase
Clonality: Recombinant
Clone: ARG1/8869R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human