Anti-EZH2 Mouse Monoclonal Antibody [clone: EZH2/6988]
Catalog # 77179-560
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:Enhancer of zeste 2
- Antigen Symbol:EZH2
- Clonality:Monoclonal
- Clone:EZH2/6988
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15910
- Antigen Synonyms:Enhancer of zeste homolog 2 (Drosophila)|enhancer of zeste 2 polycomb repressive complex 2 subunit|Enx1h|ENX1|Enhancer of zeste, Drosophila, homolog 2|Enhancer of zeste homolog 2|ENX-1|Enx 1h|ENX 1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:81 - 102 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 300-550) of human EZH2 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77179-560
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [EZH2/6988] antibody to EZH2 for ELISA, Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ ml, WB: 1 to 2 µg/ ml, IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone: EZH2/6988
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Human