Anti-ZIP4 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to ZIP4 for WB with samples derived from Oryza sativa.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: ZIP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Oryza sativa
Specifications
- Catalog No:
- 77201-982
- Antigen Symbol:
- ZIP4
- Antigen Name:
- ZIP4
- Antigen Synonyms:
- MGC156705|Acrodermatitis enteropathica zinc deficiency type|AEZ|Activated in W/Wv mouse stomach 2|1600025H15Rik|MGC74741|AWMS2|S39A4_HUMAN|AU041686|FLJ20327
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Oryza sativa
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q5N829
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant protein of Oryza sativa ZIP4.
- Form:
- Liquid
- Molecular Weight:
- 103 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C