Anti-TIM44 Rabbit Monoclonal Antibody [clone: ARC3044]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC3044] antibody to TIM44 for WB with samples derived from Human and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: TIM44
Clonality: Monoclonal
Clone: ARC3044
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77207-498
- Antigen Symbol:
- TIM44
- Antigen Name:
- DKFZp686H05241
- Antigen Synonyms:
- Mitochondrial inner membrane translocase|TIMM44|MIMT44|Translocase of inner mitochondrial membrane 44 homolog (yeast)|Translocase of inner mitochondrial membrane 44|TIM44
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC3044
- Reactivity:
- Human, Rat
- Host:
- Rabbit
- Gene ID:
- UniprotID# O43615
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant protein of Human TIM44.
- Form:
- Liquid
- Molecular Weight:
- 45 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C