Anti-SMAD5 (phospho Ser465) Rabbit Monoclonal Antibody [clone: ARC59204]
Rabbit monoclonal [ARC59204] antibody to SMAD5 (phospho Ser465) for WB with samples derived from Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:1,000 to 1:5,000
Type: Primary
Antigen: SMAD5
Clonality: Monoclonal
Clone: ARC59204
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat
- Catalog No:
- 77214-378
- Antigen Symbol:
- SMAD5
- Antigen Name:
- DKFZp781C1895
- Antigen Synonyms:
- JV5 1|hSmad5|MADH5|Dwfc|JV5-1|MAD homolog 5|DKFZp781O1323|MAD, mothers against decapentaplegic homolog 5|MADH 5
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC59204
- Reactivity:
- Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q99717
- Isotype:
- IgG
- Western Blot:
- Yes
- Modification:
- phospho Ser465
- Immunogen:
- A synthetic phosphorylated peptide around S465 of human Smad5.
- Form:
- Liquid
- Molecular Weight:
- 60 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C