About this item
Rabbit polyclonal antibody to Giantin for WB and ICC/IF with samples derived from Human.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:1,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: Giantin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77209-626
- Antigen Symbol:
- Giantin
- Antigen Name:
- 372 kDa Golgi complex associated protein
- Antigen Synonyms:
- GCP372|Golgi autoantigen golgin subfamily b macrogolgin (with transmembrane signal) 1|GOLGB1|Giantin|Golgi autoantigen, golgin subfamily b, macrogolgin 1|Golgi autoantigen golgin subfamily a member 1|GOGB1_HUMAN|GCP|372 kDa Golgi complex-associated protein
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q14789
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant protein of human Giantin.
- Form:
- Liquid
- Molecular Weight:
- 376 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.05% Proclin 300
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF