Anti-HERC4 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to HERC4 for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: HERC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77232-832
- Antigen Symbol:
- HERC4
- Antigen Name:
- DKFZP564G092
- Antigen Synonyms:
- HECT domain and RCC1-like domain-containing protein 4|HERC4_HUMAN|HERC4|KIAA1593|HERC 4|HECT and RLD domain containing E3 ubiquitin protein ligase 4|HECT domain and RCC1 like domain containing protein 4|Probable E3 ubiquitin protein ligase HERC4|Hect domain and RLD 4
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q5GLZ8
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human HERC4 (NP_056416.2).
- Sequence:
- WTVNEALIQKWLSYPSGRFPVEIANEIDGTFSSSGCLNGSFLAVSNDDHYRTGTRFSGVDMNAARLLFHKLIQPDHPQISQQVAASLEKNLIPKLTSSLPDVEALRFYLTLPECPLMSDSNNFTTIAIPFGTALVNLEKAPLKVLENWWSVLEPPLFLKIVELFKEVVVHLLKLYKIGIPPSERRIFNSFLHTALKVLEILHRVNEKMGQIIQYDKFYIHEVQELIDIRNDYINWVQQQAYGMLADIPVTICTYPFVFDAQAKTTLLQTDAVLQMQMAIDQAHRQNVSSLFLPVIESVNPC
- Form:
- Liquid
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C