Anti-ZNF202 Mouse Monoclonal Antibody [clone: PCRP-ZNF202-1C4]
Catalog # 77178-512
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# O95125
- Antigen Synonyms:ZNF 202|ZKSCAN 10|ZNF202|ZKSCAN10|Zinc finger protein with KRAB and SCAN domains 10|ZN202_HUMAN
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:74.72 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 37-132) of human ZNF202 protein
- Purification:Protein A/G chromatography
- Cat. No.:77178-512
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-ZNF202-1C4] antibody to ZNF202 for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: ZNF202
Clonality: Monoclonal
Clone: PCRP-ZNF202-1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human