Anti-TTF1 Rabbit Recombinant Antibody [clone: TTF1/8123R]
Catalog # 77178-452
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AV026640
- Antigen Symbol:TTF1
- Clonality:Recombinant
- Clone:TTF1/8123R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P43699
- Antigen Synonyms:Homeobox protein Nkx2.1|Homeobox protein Nkx 2.1|BCH|NK 2|BHC|Homeobox protein NK 2 homolog A|Homeobox protein NK-2 homolog A|Benign chorea|Homeobox protein Nkx-2.1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-400) of human NXK2-1 (TTF-1) protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77178-452
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [TTF1/8123R] antibody to TTF1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: TTF1
Clonality: Recombinant
Clone: TTF1/8123R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human