Anti-Periostin Mouse Monoclonal Antibody [clone: POSTN/3502]
77177-920
- Antibody Type:Primary
- Antigen Name:Fasciclin-I like
- Antigen Symbol:Periostin
- Clonality:Monoclonal
- Clone:POSTN/3502
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15063
- Antigen Synonyms:Osteoblast specific factor|OSF2|OSF-2|Osteoblast specific factor 2|MGC119510|Osteoblast-specific factor 2|MGC119511|OSF 2|Osteoblast specific factor 2 (fasciclin I like)
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:84 kDa/74 kDa (Periostin) kDa/90 kDa (secreted glycoprotein)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 193-326) of human POSTN protein. The exact sequence is proprietary.
- Purification:Protein A/G chromatography
- Cat. No.:77177-920
- Packaging:Plastic vial
Mouse monoclonal [POSTN/3502] antibody to Periostin for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: Periostin
Clonality: Monoclonal
Clone: POSTN/3502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human