Anti-pan HLA Class II Mouse Recombinant Antibody [clone: rHLA-Pan/8847]
Catalog # 77177-822
Supplier: ANTIBODIES.COM LLC
Specifications
- Form:Liquid
- Gene ID:UniprotID# P04440 (HLA-DP), P01908, P01909, P01920 (HLA-DQ), P01903 (HLA-DR)
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:33 - 35 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Priess human B cell line
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:77177-822
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rHLA-Pan/8847] antibody to pan HLA Class II for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: pan HLA Class II
Clonality: Recombinant
Clone: rHLA-Pan/8847
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human