Anti-ORP1 Rabbit Monoclonal Antibody [clone: ARC2772]
Supplier: Avantor
Certificates
About this item
Rabbit monoclonal [ARC2772] antibody to ORP1 for WB and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500 to 1:1,000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: ORP1
Clonality: Monoclonal
Clone: ARC2772
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat