Anti-alpha SNAP Rabbit Monoclonal Antibody [clone: ARC2382]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2382] antibody to alpha SNAP for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: alpha SNAP
Clonality: Monoclonal
Clone: ARC2382
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat