Anti-PDGF B Rabbit Monoclonal Antibody [clone: ARC53713]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC53713] antibody to PDGF B for ICC/IF with samples derived from Mouse.
- Validated Applications: ICC/IF
- Recommended Dilutions: ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: PDGF B
Clonality: Monoclonal
Clone: ARC53713
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77230-244
- Antigen Symbol:
- PDGF B
- Antigen Name:
- Becaplermin
- Antigen Synonyms:
- Oncogene SIS|PDGF subunit B|PDGF Beta|PDGF 2|FLJ12858|c sis|PDGF B chain|PDGF B|PDGF-2
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC53713
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P01127
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 142-241 of human PDGFB (NP_002599.1)
- Sequence:
- RPTQVQLRPVQVRKIEIVRKKPIFKKATVTLEDHLACKCETVAAARPVTRSPGGSQEQRAKTPQTRVTIRTVRVRRPPKGKHRKFKHTHDKTALKETLGA
- Form:
- Liquid
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.05% Proclin 300
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- ICC/IF