Anti-EEFSEC Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to EEFSEC for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: EEFSEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human