Anti-NPAS3 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to NPAS3 for WB with samples derived from Human and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: NPAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat