Anti-AKT1 +AKT2 + AKT3 (phospho Ser473) Rabbit Monoclonal Antibody [clone: ARC5023-06]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC5023-06] antibody to AKT1 +AKT2 + AKT3 (phospho Ser473) for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: AKT1 +AKT2 + AKT3
Clonality: Monoclonal
Clone: ARC5023-06
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat