Anti-Cytochrome P450 4A/CYP4A11 Rabbit Monoclonal Antibody [clone: ARC2219]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2219] antibody to Cytochrome P450 4A/CYP4A11 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: Cytochrome P450 4A/CYP4A11
Clonality: Monoclonal
Clone: ARC2219
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat