Anti-OFD1 Rabbit Polyclonal Antibody
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit polyclonal antibody to OFD1 for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: OFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77229-192
- Antigen Symbol:
- OFD1
- Antigen Name:
- 71 7A
- Antigen Synonyms:
- JBTS10|Protein 71-7A|Oral-facial-digital syndrome 1 protein|OTTHUMP00000022941|717A|OFD1|OFD1_HUMAN|CXorf5|oral facial digital syndrome 1
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# O75665
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 763-1012 of human OFD1 (NP_003602.1)
- Sequence:
- PSPCPDRMPLPSPTESRHSLSIPPVSSPPEQKVGLYRRQTELQDKSEFSDVDKLAFKDNEEFESSFESAGNMPRQLEMGGLSPAGDMSHVDAAAAAVPLSYQHPSVDQKQIEEQKEEEKIREQQVKERRQREERRQSNLQEVLERERRELEKLYQERKMIEESLKIKIKKELEMENELEMSNQEIKDKSAHSENPLEKYMKIIQQEQDQESADKSSKKMVQEGSLVDTLQSSDKVESLTGFSHEELDDSW
- Form:
- Liquid
- Molecular Weight:
- 117 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C