Anti-NPR-B Rabbit Monoclonal Antibody [clone: ARC2421]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC2421] antibody to NPR-B for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: NPR-B
Clonality: Monoclonal
Clone: ARC2421
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77228-460
- Antigen Symbol:
- NPR-B
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC2421
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P20594
- Isotype:
- IgG
- Western Blot:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 800-900 of human ANPRB/NRP2 (P20594)
- Sequence:
- NLLLRMEQYANNLEKLVEERTQAYLEEKRKAEALLYQILPHSVAEQLKRGETVQAEAFDSVTIYFSDIVGFTALSAESTPMQVVTLLNDLYTCFDAIIDNF
- Form:
- Liquid
- Molecular Weight:
- 130 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C