Rabbit monoclonal [ARC1572] antibody to Src (phospho Tyr419) for WB with samples derived from Human and Mouse.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Src
Clonality: Monoclonal
Clone: ARC1572
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77228-340
- Antigen Symbol:
- Src
- Antigen Name:
- ASV
- Antigen Synonyms:
- c SRC|Oncogene SRC|Neuronal SRC|CDNA FLJ14219 fis clone NT2RP3003800 highly similar to Rattus norvegicus tyrosine protein kinase pp60 c src mRNA|Neuronal CSRC tyrosine specific protein kinase|OTTHUMP00000174476|Avian sarcoma virus|EC 2.7.10.2|cSrc
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1572
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P12931
- Isotype:
- IgG
- Western Blot:
- Yes
- Modification:
- phospho Tyr419
- Immunogen:
- A synthetic phosphorylated peptide around Y419 of human Phospho-Src-Y419 (P12931)
- Sequence:
- NEYTA
- Form:
- Liquid
- Molecular Weight:
- 60 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C