Anti-IQGAP1 Rabbit Monoclonal Antibody [clone: ARC1739]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1739] antibody to IQGAP1 for WB and IHC with samples derived from Human and Mouse.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200
Type: Primary
Antigen: IQGAP1
Clonality: Monoclonal
Clone: ARC1739
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77227-048
- Antigen Symbol:
- IQGAP1
- Antigen Name:
- HUMORFA01
- Antigen Synonyms:
- p195|IQGA1_HUMAN|IQGAP1|KIAA0051|Ras GTPase activating like protein 1|Ras GTPase-activating-like protein IQGAP1|RasGAP-like with IQ motifs|SAR1|IQ motif containing GTPase activating protein 1
- Antibody Type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- ARC1739
- Reactivity:
- Human, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# P46940
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- A synthetic peptide corresponding to a sequence within amino acids 1-100 of human IQGAP1 (P46940)
- Sequence:
- MSAADEVDGLGVARPHYGSVLDNERLTAEEMDERRRQNVAYEYLCHLEEAKRWMEACLGEDLPPTTELEEGLRNGVYLAKLGNFFSPKVVSLKKIYDREQ
- Form:
- Liquid
- Molecular Weight:
- 195 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol, 0.05% BSA, and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- IHC