To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Liver X receptor alpha
- Antigen Symbol:LXR alpha + LXR beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Antigen Synonyms:NR1H3|LXRbeta|LXRalpha|Nuclear orphan receptor LXR alpha|LXR b|NR1H2|Liver X receptor beta|LXR a|LXR alpha + LXR beta
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:CRLQDKKLPPLLSEI
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human NR1H3; NR1H2 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77072-780
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to LXR alpha + LXR beta for ELISA, WB, IF and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, WB, IF, Flow Cytometry
Type: Primary
Antigen: LXR alpha + LXR beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human